Sodium ((1S,4R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonate 98%

Supplier: Ambeed
Warning

A947802-1000G A947802-100G A947802-25G A947802-500G A947802-5G
76698-808EA 252.5 USD
76698-808 76698-810 76698-812 76698-814 76698-816
Sodium ((1S,4R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonate 98%
Sodium ((1S,4R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonate

Precaución: for Research Use Only

Formula: C₁₀H₁₅NaO₄S
MW: 254.27 g/mol
MDL Number: MFCD00216827
CAS Number: 21791-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR