(R)-4-Phenylthiazolidine-2-thione 98%

Supplier: Ambeed
Warning

A943278-1G A943278-250MG
76698-788EA 55.17 USD
76698-788 76698-790
(R)-4-Phenylthiazolidine-2-thione 98%
(R)-4-Phenylthiazolidine-2-thione

Precaución: for Research Use Only

Formula: C₉H₉NS₂
MW: 195.30 g/mol
MDL Number: MFCD06658215
CAS Number: 110199-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR