Allyl cyclohexanepropionate 98%

Supplier: Ambeed
Warning

Synonyms: Allyl Cyclohexanepropionate, Cyclohexanepropanoic acid 2-propen-1-yl ester, Allyl 3-cyclohexylpropanoate, 2-Propenyl 3-cyclohexylpropanoate, Cyclohexanepropionic acid allyl ester, Allyl 3-cyclohexylpropionate, 3-Cyclohexylpropionic acid Allyl Ester Cyclohexanepropionic acid Allyl Ester

A941368-100G A941368-25G A941368-500G
76698-782EA 9.55 USD
76698-782 76698-784 76698-786
Allyl cyclohexanepropionate 98%
Allyl cyclohexanepropionate

Precaución: for Research Use Only

Formula: C₁₂H₂₀O₂
MW: 196.29 g/mol
MDL Number: MFCD00059557
CAS Number: 2705-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR