Ethyl-cis-4,7,10,13,16,19-docosahexaenoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl docosa-4,7,10,13,16,19-hexaenoate, cis-4,7,10,13,16,19-Docosahexaenoic Acid Ethyl Ester, DHA Ethyl Ester

A940171-100MG A940171-250MG A940171-5G A940171-1G
76698-776EA 20.16 USD
76698-776 76698-778 76698-780 77603-324
Ethyl-cis-4,7,10,13,16,19-docosahexaenoate 98%
Ethyl-cis-4,7,10,13,16,19-docosahexaenoate

Precaución: for Research Use Only

Formula: C₂₄H₃₆O₂
MW: 356.55 g/mol
MDL Number: MFCD00056293
CAS Number: 84494-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR