Sodium-3-(N-ethylanilino)propanesulfonate 98+%

Supplier: Ambeed
Warning

Synonyms: N-Ethyl-N-(3-sulphopropyl)aniline Sodium Salt, ALPS, 3-(N-Ethylanilino)propanesulphonic acid Sodium Salt, Sodium 3-(ethyl(phenyl)amino)propane-1-sulphonate

A938719-1G A938719-250MG A938719-5G
76698-766EA 55 USD
76698-766 77627-768 77627-770
Sodium-3-(N-ethylanilino)propanesulfonate 98+%
Sodium-3-(N-ethylanilino)propanesulfonate

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₃S
MDL Number: MFCD01311047
CAS Number: 82611-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR