(R)-Methyl 4-(1-aminoethyl)benzoate 98%

Supplier: Ambeed
Warning

A936664-100MG A936664-1G A936664-250MG A936664-5G A936664-25G
76698-756EA 7 USD
76698-756 76698-758 76698-760 76698-762 77878-747
(R)-Methyl 4-(1-aminoethyl)benzoate 98%
(R)-Methyl 4-(1-aminoethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.216 g/mol
MDL Number: MFCD06762071
CAS Number: 912342-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR