8-Chloro-3-methyl[1,2,4]triazolo[4,3-a]pyrazine 98%

Supplier: Ambeed
Warning

Synonyms: 8-Chloro-3-methyl-[1,2,4]triazolo[4,3-a]pyrazine, 8-Chloro-3-methyltriazolo[4,3-a]pyrazine

A934528-10G A934528-1G A934528-250MG A934528-5G
76698-748EA 404.21 USD
76698-748 76698-750 76698-752 76698-754
8-Chloro-3-methyl[1,2,4]triazolo[4,3-a]pyrazine 98%
8-Chloro-3-methyl[1,2,4]triazolo[4,3-a]pyrazine

Precaución: for Research Use Only

Formula: C₆H₅ClN₄
MW: 168.584 g/mol
MDL Number: MFCD08272087
CAS Number: 68774-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR