1-Phenyl-2-pyrrolidone 97%

Supplier: Ambeed
Warning

Synonyms: 1-Phenyl-2-pyrrolidinone

A931673-100G A931673-10G A931673-25G A931673-5G A931673-500G
76698-734EA 82 USD
76698-734 76698-736 76698-738 76698-740 77627-764
1-Phenyl-2-pyrrolidone 97%
1-Phenyl-2-pyrrolidone

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO
MDL Number: MFCD00003192
CAS Number: 4641-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR