D-γ-Tocopherol 98%

Supplier: Ambeed
Warning

Synonyms: (R)-2,7,8-Trimethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-ol, γ-Tocopherol

A929586-100MG A929586-25MG A929586-250MG
76698-724EA 201.58 USD
76698-724 76698-726 77627-762
D-γ-Tocopherol 98%
D-γ-Tocopherol

Precaución: for Research Use Only

Formula: C₂₈H₄₈O₂
MW: 416.69 g/mol
MDL Number: MFCD00066529
CAS Number: 54-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR