4-Fluoro-3-methylbenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (4-Fluoro-3-methylphenyl)methanol

A919760-100MG A919760-1G A919760-250MG
76698-694EA 23.34 USD
76698-694 76698-696 76698-698
4-Fluoro-3-methylbenzyl alcohol 98%
4-Fluoro-3-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉FO
MW: 140.16 g/mol
MDL Number: MFCD01631427
CAS Number: 261951-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR