2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-β-D-glucopyranose 98%
Supplier: Ambeed
|
Warning
|
A915516-1G
A915516-250MG
A915516-5G
A915516-100G
A915516-25G
76698-686EA
7
USD
76698-686
76698-688
76698-690
77627-750
77878-738
2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-β-D-glucopyranose 98%
2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-beta-D-glucopyranose
Precaución: for Research Use Only
Formula:
C₁₆H₂₃NO₁₀ |
MDL Number:
MFCD00006595 CAS Number: 7772-79-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...