Potassium hexadecyl hydrogenphosphate 97%

Supplier: Ambeed
Danger

A910049-1G A910049-5G A910049-10G
76698-662EA 22.28 USD
76698-662 76698-664 77627-736
Potassium hexadecyl hydrogenphosphate 97%
Potassium hexadecyl hydrogenphosphate

Precaución: for Research Use Only

Formula: C₁₆H₃₄KO₄P
MW: 360.51 g/mol
MDL Number: MFCD04112600
CAS Number: 19035-79-1
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR