3-Fluoro-2-methoxyphenylboronic acid 95%
Supplier: Ambeed
|
Warning
|
A909835-100MG
A909835-1G
A909835-250MG
A909835-25G
A909835-5G
76698-652EA
7
USD
76698-652
76698-654
76698-656
76698-658
76698-660
3-Fluoro-2-methoxyphenylboronic acid 95%
3-Fluoro-2-methoxyphenylboronic acid
Precaución: for Research Use Only
Formula:
C₇H₈BFO₃ |
MDL Number:
MFCD03095355 CAS Number: 762287-59-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...