Cyclohexyl(dimethoxy)methylsilane 98%

Supplier: Ambeed
Danger

A907305-25G A907305-500G
76698-636EA 5 USD
76698-636 77880-733
Cyclohexyl(dimethoxy)methylsilane 98%
Cyclohexyl(dimethoxy)methylsilane

Precaución: for Research Use Only

Formula: C₉H₂₀O₂Si
MDL Number: MFCD00071752
CAS Number: 17865-32-6
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR