9-Decen-1-ol 97% stabilized
Supplier: Ambeed
|
Warning
|
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC)
A906722-100G
A906722-500G
A906722-25G
A906722-5G
76698-632EA
54
USD
76698-632
76698-634
77603-266
77878-724
9-Decen-1-ol 97% stabilized
9-Decen-1-ol
Precaución: for Research Use Only
Formula:
C₁₀H₂₀O |
MDL Number:
MFCD00002992 CAS Number: 13019-22-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...