1-(Benzyloxycarbonyl)homopiperazine 95%

Supplier: Ambeed
Warning

Synonyms: Benzyl 1,4-diazepane-1-carboxylate

A905737-1G A905737-250MG A905737-25G A905737-5G
76698-624EA 65.78 USD
76698-624 76698-626 76698-628 76698-630
1-(Benzyloxycarbonyl)homopiperazine 95%
1-(Benzyloxycarbonyl)homopiperazine

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₂
MW: 234.2942 g/mol
MDL Number: MFCD01863703
CAS Number: 117009-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR