3-Fluoro-4-methoxybenzyl bromide 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-(Bromomethyl)-2-fluoro-1-methoxybenzene
A900829-100MG
A900829-10G
A900829-1G
A900829-250MG
A900829-5G
A900829-25G
76698-604EA
5.31
USD
76698-604
76698-606
76698-608
76698-610
76698-612
77595-450
3-Fluoro-4-methoxybenzyl bromide 97%
3-Fluoro-4-methoxybenzyl bromide
Precaución: for Research Use Only
Formula:
C₈H₈BrFO MW: 219.05 g/mol |
MDL Number:
MFCD00671768 CAS Number: 331-61-3 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...