Cresyl violet (acetate) 65%

Supplier: Ambeed
Warning

A900422-1G A900422-5G A900422-10G
76698-600EA 15 USD
76698-600 76698-602 77627-720
Cresyl violet (acetate) 65%
Cresyl violet (acetate)

Precaución: for Research Use Only

Formula: C₁₈H₁₅N₃O₃
MDL Number: MFCD00013151
CAS Number: 10510-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR