Benzyl cyanoacetate 98%

Supplier: Ambeed
Warning

A900144-1G A900144-25G A900144-5G A900144-100G
76698-594EA 9 USD
76698-594 76698-596 76698-598 77627-718
Benzyl cyanoacetate 98%
Benzyl cyanoacetate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MDL Number: MFCD00051719
CAS Number: 14447-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR