Calcon® (mordant black 17) 70+%

Supplier: Ambeed
Warning

Synonyms: Palatine chrome black 6BN, Mordant Black 17, Eriochrome® Blue Black R, Sodium 3-hydroxy-4-((2-hydroxynaphthalen-1-yl)diazenyl)naphthalene-1-sulphonate, Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-1-naphthalenesulphonate

A898143-1000G A898143-100G A898143-25G A898143-500G
76698-578EA 87 USD
76698-578 76698-580 76698-582 76698-584
Calcon® (mordant black 17) 70+%
Calcon® (mordant black 17)

Precaución: for Research Use Only

Formula: C₂₀H₁₃N₂NaO₅S
MW: 416.38 g/mol
MDL Number: MFCD00004069
CAS Number: 2538-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR