Azobenzene-4,4'-dicarboxylic acid 95% (by HPLC)
Supplier: Ambeed
|
Warning
|
Synonyms:
Azobenzene-4,4'-dicarboxylic Acid, 4,4'-Azodibenzoic acid, 4,4'-Azodibenzoic Acid, 4,4'-(Diazene-1,2-diyl)dibenzoic acid
A896211-1G
A896211-250MG
A896211-5G
76698-572EA
39.26
USD
76698-572
76698-574
76698-576
Azobenzene-4,4'-dicarboxylic acid 95% (by HPLC)
Azobenzene-4,4'-dicarboxylic acid
Precaución: for Research Use Only
Formula:
C₁₄H₁₀N₂O₄ MW: 270.24 g/mol |
MDL Number:
MFCD00496621 CAS Number: 586-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...