3,4-Ethylenedioxynitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 6-Nitro-2,3-dihydrobenzo[b][1,4]dioxine, 6-Nitro-1,4-benzodioxane

A895378-1G A895378-5G A895378-25G
76698-564EA 7 USD
76698-564 77627-716 77878-720
3,4-Ethylenedioxynitrobenzene 98%
3,4-Ethylenedioxynitrobenzene

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MW: 181.15 g/mol
MDL Number: MFCD00088689
CAS Number: 16498-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR