cis-3-Hexenyl isobutyrate 95%

Supplier: Ambeed
Warning

Synonyms: (Z)-Hex-3-en-1-yl isobutyrate, Isobutyric Acid cis-3-Hexenyl Ester

A892151-100G A892151-25G
76698-560EA 46 USD
76698-560 76698-562
cis-3-Hexenyl isobutyrate 95%
cis-3-Hexenyl isobutyrate

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₂
MDL Number: MFCD00036529
CAS Number: 41519-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR