Bis(4-tert-butylphenyl)iodonium trifluoromethanesulfonate 98%

Supplier: Ambeed
Danger

Synonyms: Bis(4-(tert-butyl)phenyl)iodonium trifluoromethanesulphonate, Bis(4-tert-butylphenyl)iodonium Triflate

A887593-100MG A887593-250MG A887593-25G A887593-5G A887593-1G
76698-534EA 7 USD
76698-534 76698-536 76698-538 76698-540 77878-718
Bis(4-tert-butylphenyl)iodonium trifluoromethanesulfonate 98%
Bis(4-tert-butylphenyl)iodonium trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₂₁H₂₆F₃IO₃S
MDL Number: MFCD02683471
CAS Number: 84563-54-2
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR