N-Benzoyl-L-phenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: Bz-Phe-OH

A884599-100G A884599-10G A884599-25G A884599-5G
76698-526EA 103.97 USD
76698-526 76698-528 76698-530 76698-532
N-Benzoyl-L-phenylalanine 98%
N-Benzoyl-L-phenylalanine

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO₃
MW: 269.3 g/mol
MDL Number: MFCD00063153
CAS Number: 2566-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR