Allyltriphenylphosphonium bromide 98%

Supplier: Ambeed
Warning

A883815-100G A883815-25G A883815-500G A883815-5G
76698-518EA 44 USD
76698-518 76698-520 76698-522 76698-524
Allyltriphenylphosphonium bromide 98%
Allyltriphenylphosphonium bromide

Precaución: for Research Use Only

Formula: C₂₁H₂₀BrP
MW: 383.261 g/mol
MDL Number: MFCD00011808
CAS Number: 1560-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR