Bis(2-morpholinoethyl)ether 97%

Supplier: Ambeed
Warning

Synonyms: 4,4'-(Oxybis(ethane-2,1-diyl))dimorpholine, 4,4'-(3-Oxapentane-1,5-diyl)bismorpholine

A882057-100G A882057-25G A882057-5G
76698-512EA 41.38 USD
76698-512 76698-514 76698-516
Bis(2-morpholinoethyl)ether 97%
Bis(2-morpholinoethyl)ether

Precaución: for Research Use Only

Formula: C₁₂H₂₄N₂O₃
MW: 244.34 g/mol
MDL Number: MFCD00072740
CAS Number: 6425-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR