Acetohydrazide 97%

Supplier: Ambeed
Danger

Synonyms: Acetic hydrazide, Acethydrazide

A879664-25G A879664-5G
76698-500EA 6.37 USD
76698-500 76698-502
Acetohydrazide 97%
Acetohydrazide

Precaución: for Research Use Only

Formula: C₂H₆N₂O
MW: 74.08 g/mol
MDL Number: MFCD00007610
CAS Number: 1068-57-1
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR