ITSA-1 99+%

Supplier: Ambeed
Warning

Synonyms: (1H-Benzo[d][1,2,3]triazol-1-yl)(2,4-dichlorophenyl)methanone

A879167-100MG A879167-10MG A879167-1MG A879167-250MG A879167-25MG A879167-50MG
76698-488EA 96.55 USD
76698-488 76698-490 76698-492 76698-494 76698-496 76698-498
ITSA-1 99+%
ITSA-1

Precaución: for Research Use Only

Formula: C₁₃H₇Cl₂N₃O
MW: 292.1202 g/mol
MDL Number: MFCD00195103
CAS Number: 200626-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR