CAPSO sodium salt (3-(cyclohexylamino)-2-hydroxypropanesulfonic acid sodium salt) 98%

Supplier: Ambeed
Warning

Synonyms: Sodium 3-(cyclohexylamino)-2-hydroxypropane-1-sulphonate

A877057-100G A877057-25G
76698-480EA 52 USD
76698-480 76698-482
CAPSO sodium salt (3-(cyclohexylamino)-2-hydroxypropanesulfonic acid sodium salt) 98%
CAPSO sodium salt (3-(Cyclohexylamino)-2-hydroxypropanesulphonic acid sodium salt)

Precaución: for Research Use Only

Formula: C₉H₁₈NNaO₄S
MDL Number: MFCD00070063
CAS Number: 102601-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR