D-(+)-Melibiose monohydrate 99%

Supplier: Ambeed
Warning

Synonyms: 6-O-α-D-Galactopyranosyl-D-glucopyranose monohydrate, (2R,3S,4R,5R)-2,3,4,5-Tetrahydroxy-6-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, alpha-D-(+)-Melibiose monohydrate

A876382-1G A876382-25G A876382-5G
76698-468EA 22 USD
76698-468 76698-470 76698-472
D-(+)-Melibiose monohydrate 99%
D-(+)-Melibiose monohydrate

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₁₁·H₂O
MW: 360.312 g/mol
MDL Number: MFCD00198188
CAS Number: 66009-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR