3-Fluoro-5-(trifluoromethyl)benzylamine 98%

Supplier: Ambeed
Danger

Synonyms: (3-Fluoro-5-(trifluoromethyl)phenyl)methanamine

A874690-1G A874690-5G
76698-464EA 13.8 USD
76698-464 76698-466
3-Fluoro-5-(trifluoromethyl)benzylamine 98%
3-Fluoro-5-(trifluoromethyl)benzylamine

Precaución: for Research Use Only

Formula: C₈H₇F₄N
MW: 193.14 g/mol
MDL Number: MFCD00061163
CAS Number: 150517-77-4
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR