Cyclopropyl phenyl sulfide 98%

Supplier: Ambeed
Warning

A874636-25G A874636-5G A874636-1G
76698-462EA 212 USD
76698-462 77595-412 77880-692
Cyclopropyl phenyl sulfide 98%
Cyclopropyl phenyl sulphide

Precaución: for Research Use Only

Formula: C₉H₁₀S
MDL Number: MFCD00009684
CAS Number: 14633-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR