Acetylacetonatobis(ethylene)rhodium(I) 98%

Supplier: Ambeed
Warning

A872555-50MG A872555-1G A872555-250MG A872555-100MG
76698-460EA 60 USD
76698-460 77603-182 77603-184 77627-696
Acetylacetonatobis(ethylene)rhodium(I) 98%
Acetylacetonatobis(ethylene)rhodium(I)

Precaución: for Research Use Only

Formula: C₉H₁₆O₂Rh
MW: 259.13 g/mol
MDL Number: MFCD00015354
CAS Number: 12082-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR