(S)-3-(Cbz-amino)-2-oxoazepane 97%

Supplier: Ambeed
Warning

A872475-1G A872475-250MG A872475-5G
76698-454EA 46.68 USD
76698-454 76698-456 76698-458
(S)-3-(Cbz-amino)-2-oxoazepane 97%
(S)-3-(Cbz-amino)-2-oxoazepane

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O₃
MW: 262.30 g/mol
MDL Number: MFCD08060107
CAS Number: 103478-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR