Methyl (3-methoxyphenyl)acetate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 3-Methoxyphenylacetate
A868290-100G
A868290-10G
A868290-1G
A868290-25G
A868290-5G
76698-444EA
287.51
USD
76698-444
76698-446
76698-448
76698-450
76698-452
Methyl (3-methoxyphenyl)acetate 98%
Methyl (3-methoxyphenyl)acetate
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₃ MW: 180.23 g/mol |
MDL Number:
MFCD00017205 CAS Number: 18927-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...