Methyl-3,5-dimethoxybenzoate 97%
Supplier: Ambeed
|
Warning
|
A860886-100G
A860886-10G
A860886-25G
A860886-500G
A860886-5G
76698-424EA
43
USD
76698-424
76698-426
76698-428
76698-430
76698-432
Methyl-3,5-dimethoxybenzoate 97%
Methyl-3,5-dimethoxybenzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₄ |
MDL Number:
MFCD00008432 CAS Number: 2150-37-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...