Cyclohexyl methacrylate 98% stabilised

Supplier: Ambeed
Danger

Synonyms: Methacrylic Acid Cyclohexyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A857998-100G A857998-25G A857998-500G
76698-414EA 28 USD
76698-414 76698-416 76698-418
Cyclohexyl methacrylate 98% stabilised
Cyclohexyl methacrylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₂
MW: 168.24 g/mol
MDL Number: MFCD00014292
CAS Number: 101-43-9
UN: 3082
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR