(2R,3R,4S,5S,6R)-2-[(2S,3S,4R,5R)-4-Hydroxy-2,5-bis(hydroxymethyl)-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxolan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol 99+%

Supplier: Ambeed
Warning

A854343-100MG A854343-1MG A854343-250MG
76698-398EA 44.56 USD
76698-398 76698-400 76698-402
(2R,3R,4S,5S,6R)-2-[(2S,3S,4R,5R)-4-Hydroxy-2,5-bis(hydroxymethyl)-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxolan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol 99+%
(2R,3R,4S,5S,6R)-2-[(2S,3S,4R,5R)-4-Hydroxy-2,5-bis(hydroxymethyl)-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxolan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol

Precaución: for Research Use Only

Formula: C₁₈H₃₂O₁₆
MW: 504.43 g/mol
MDL Number: MFCD00006627
CAS Number: 597-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR