Benzyltributylammonium chloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Benzyl-N,N-dibutylbutan-1-aminium chloride
A847514-1000G
A847514-100G
A847514-10G
A847514-25G
A847514-500G
76698-376EA
64
USD
76698-376
76698-378
76698-380
76698-382
76698-384
Benzyltributylammonium chloride 97%
Benzyltributylammonium chloride
Precaución: for Research Use Only
Formula:
C₁₉H₃₄ClN MW: 311.94 g/mol |
MDL Number:
MFCD00011849 CAS Number: 23616-79-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...