Sodium benzenesulfinate dihydrate 98%

Supplier: Ambeed
Warning

A846536-1000G A846536-100G A846536-25G A846536-500G
76698-362EA 60 USD
76698-362 76698-364 76698-366 76698-368
Sodium benzenesulfinate dihydrate 98%
Sodium benzenesulphinate dihydrate

Precaución: for Research Use Only

Formula: C₆H₅NaO₂S·2H₂O
MDL Number: MFCD00149638
CAS Number: 25932-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR