Lumefantrine 98%

Supplier: Ambeed
Warning

Synonyms: (9Z)-2,7-Dichloro-9-[(4-chlorophenyl)methylene]-alpha-[(dibutylamino)methyl]-9H-fluoren-4-methanol, Benflumetol, (Z)-2-(Dibutylamino)-1-(2,7-dichloro-9-(4-chlorobenzylidene)-9H-fluoren-4-yl)ethanol

A846348-100MG A846348-1G A846348-25G A846348-5G A846348-100G A846348-500G
76698-354EA 5.31 USD
76698-354 76698-356 76698-358 76698-360 77603-146 77603-148
Lumefantrine 98%
Lumefantrine

Precaución: for Research Use Only

Formula: C₃₀H₃₂Cl₃NO
MW: 528.94 g/mol
MDL Number: MFCD05662268
CAS Number: 82186-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR