4-(Dimethylamino)benzylamine 97%

Supplier: Ambeed
Danger

Synonyms: 4-(Aminomethyl)-N,N-dimethylaniline

A844192-1G A844192-25G A844192-5G
76698-340EA 16 USD
76698-340 77627-646 77878-691
4-(Dimethylamino)benzylamine 97%
4-(Dimethylamino)benzylamine

Precaución: for Research Use Only

Formula: C₉H₁₄N₂
MDL Number: MFCD00466364
CAS Number: 19293-58-4
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR