Cyclohexene-1-carbonitrile 97%

Supplier: Ambeed
Warning

A840816-1G A840816-250MG A840816-5G
76698-316EA 43 USD
76698-316 76698-318 76698-320
Cyclohexene-1-carbonitrile 97%
Cyclohexene-1-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₉N
MDL Number: MFCD00013772
CAS Number: 1855-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR