2-Phenoxyethyl acetate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Acetic Acid 2-Phenoxyethyl Ester, Ethylene Glycol Monophenyl Ether Acetate
A835879-1000G
A835879-100G
A835879-25G
A835879-500G
76698-294EA
78
USD
76698-294
76698-296
76698-298
76698-300
2-Phenoxyethyl acetate 98%
2-Phenoxyethyl acetate
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₃ MW: 180.20 g/mol |
MDL Number:
MFCD00059336 CAS Number: 6192-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...