1,2-Phenylenedioxydiacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: O-Phenylenedioxydiacetic Acid, Catechol-O,O-diacetic acid, 2,2-(1,2-Phenylenebis(oxy))diacetic acid

A835755-1G A835755-25G A835755-5G
76698-288EA 15.92 USD
76698-288 76698-290 76698-292
1,2-Phenylenedioxydiacetic acid 98%
1,2-Phenylenedioxydiacetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₆
MW: 226.19 g/mol
MDL Number: MFCD00014353
CAS Number: 5411-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR