1,3-Dibenzyloxybenzene 98%

Supplier: Ambeed
Warning

Synonyms: 1,3-Bis(benzyloxy)benzene

A835561-1G A835561-25G A835561-5G
76698-282EA 11.67 USD
76698-282 76698-284 76698-286
1,3-Dibenzyloxybenzene 98%
1,3-Dibenzyloxybenzene

Precaución: for Research Use Only

Formula: C₂₀H₁₈O₂
MW: 290.3557 g/mol
MDL Number: MFCD00690156
CAS Number: 3769-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR