2,5-Dimethylbenzonitrile 97%

Supplier: Ambeed
Warning

A834613-1G A834613-250MG A834613-25G A834613-5G
76698-274EA 28.65 USD
76698-274 76698-276 76698-278 76698-280
2,5-Dimethylbenzonitrile 97%
2,5-Dimethylbenzonitrile

Precaución: for Research Use Only

Formula: C₉H₉N
MW: 131.18 g/mol
MDL Number: MFCD00001793
CAS Number: 13730-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR