2-Hydroxy-4'-methoxyacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-1-(4-methoxyphenyl)-1-ethanone, 2-Hydroxy-1-(4-methoxyphenyl)ethanone, α-Hydroxy-4'-methoxyacetophenone

A833072-100MG A833072-250MG
76698-264EA 42.44 USD
76698-264 77627-636
2-Hydroxy-4'-methoxyacetophenone 97%
2-Hydroxy-4'-methoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD00004668
CAS Number: 4136-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR