4-Oxo-3,4-dihydroquinazoline-8-carboxylic acid 95%

Supplier: Ambeed
Warning

A832577-100MG A832577-250MG
76698-260EA 55 USD
76698-260 76698-262
4-Oxo-3,4-dihydroquinazoline-8-carboxylic acid 95%
4-Oxo-3,4-dihydroquinazoline-8-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆N₂O₃
MDL Number: MFCD10566726
CAS Number: 19181-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR